Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APOBEC3D Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297987F
Description
APOBEC3D Polyclonal specifically detects APOBEC3D in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
APOBEC3D | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
APOBEC3DE, APOBEC3E, apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3D, apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3D (putative), apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3E pseudogene, ARP6, DNA dC->dU-editing enzyme APOBEC-3D, EC 3.5.4, EC 3.5.4.-, probable DNA dC->dU-editing enzyme APOBEC-3D | |
Produced in rabbits immunized with E. coli-derived Human APOBEC3D fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
FITC | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
140564 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction