Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APOBEC3D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563066
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563066 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563066 Supplier Invitrogen™ Supplier No. PA563066
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VFRGPVLPKR QSNHRQEVYF RFENHAEMCF Highest antigen sequence identity to the following orthologs: Mouse - 37%, Rat - 33%.

This gene is a member of the cytidine deaminase gene family. It is one of a group of related genes found in a cluster, thought to result from gene duplication, on chromosome 22. Members of the cluster encode proteins that are structurally and functionally related to the C to U RNA-editing cytidine deaminase APOBEC1 and inhibit retroviruses, such as HIV, by deaminating cytosine residues in nascent retroviral cDNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APOBEC3D
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene APOBEC3D
Gene Accession No. Q96AK3
Gene Alias A3D; APOBEC3D; APOBEC3DE; APOBEC3E; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3D; apolipoprotein B mRNA editing enzyme catalytic subunit 3D; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3D; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3E pseudogene; ARP6; DNA dC->dU-editing enzyme APOBEC-3D; probable DNA dC->dU-editing enzyme APOBEC-3D
Gene Symbols APOBEC3D
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human APOBEC3D. Recombinant protein control fragment (Product #RP-101628).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140564
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.