Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ APOBEC3F Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113117
Description
APOBEC3F Polyclonal Antibody for Western Blot, ICC/IF, ELISA
This protein is a member of the cytidine deaminase gene family. It is one of seven related genes or pseudogenes found in a cluster, thought to result from gene duplication, on chromosome 22. Members of the cluster encode proteins that are structurally and functionally related to the C to U RNA-editing cytidine deaminase APOBEC1. It is thought that the proteins may be RNA editing enzymes and have roles in growth or cell cycle control.
Specifications
| APOBEC3F | |
| Polyclonal | |
| Unconjugated | |
| APOBEC3F | |
| A3F; APOBEC3F; apolipoprotein B editing enzyme catalytic polypeptide-like 3F; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3F; apolipoprotein B mRNA editing enzyme catalytic subunit 3F; apolipoprotein B mRNA editing enzyme cytidine deaminase; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3F; apolipoprotein B mRNA-editing enzyme catalytic polypeptide-like 3F; ARP8; BK150C2.4.MRNA; DNA dC->dU-editing enzyme APOBEC-3F; induced upon T-cell activation; KA6 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 200316 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8IUX4 | |
| APOBEC3F | |
| Recombinant Human DNA dC->dU-editing enzyme APOBEC-3F protein (139-287AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction