Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APOBEC3G Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$610.00
Specifications
Antigen | APOBEC3G |
---|---|
Concentration | 0.05mg/mL |
Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Description
APOBEC3G Polyclonal specifically detects APOBEC3G in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
APOBEC3G | |
Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Human | |
APOBEC-related cytidine deaminase, APOBEC-related protein, APOBEC-related protein 9, apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3G, ARCD, ARP-9, bK150C2.7, CEM-15, CEM15ARP9, dJ494G10.1, DNA dC->dU-editing enzyme APOBEC-3G, EC 3.5.4, EC 3.5.4.-, EC 3.5.4.5, FLJ12740DNA dC->dU editing enzyme, MDS019, phorbolin-like protein MDS019 | |
APOBEC3G | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
0.05mg/mL | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
60489 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:MHNDTWVLLNQRRGFLCNQAPHKHGFLEGRHAELCFLDVIPFWKLDLDQDYRVTCFTSWSPCFSCAQEMAKFISKNKHVSLCIFTARIYDDQGRCQEGLRTLAEAGAKISIMTYSEFKHCWDTFVDHQGCPFQPWDGLDEHSQDLSGRL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title