Learn More
Invitrogen™ APOBEC3G Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110643
Description
Immunogen sequence: MHNDTWVLLN QRRGFLCNQA PHKHGFLEGR HAELCFLDVI PFWKLDLDQD YRVTCFTSWS PCFSCAQEMA KFISKNKHVS LCIFTARIYD DQGRCQEGLR TLAEAGAKIS IMTYSEFKHC WDTFVDHQGC PFQPWDGLDE HSQDLSGRL Highest antigen sequence indentity to the following orthologs: Mouse - 39%, Rat - 39%.
The Apoliprotein B mRNA-editing, enzyme-catalytic, polypeptide-like (APOBEC) 3 is a multi-isoform member of the cytidine deaminase family of enzymes that act on monomeric nucleoside and nucleotide substrates. Similar to TRIM5-a which targets incoming retroviral capsids, APOBEC3 plays a major role in cellular defense against retroviral infection as at least two isoforms, APOBEC3G and to a lesser extent APOBEC3F, can be incorporated HIV-1 virions and induce hypermutation in the newly synthesized viral DNA and thus destabilize the viral genome. This innate mechanism of retroviral resistance is counteracted by the HIV-1 Vif protein by inducing the ubiquitization and degradation of APOBEC3G; a single amino acid substitution (D128K) blocks APOBEC3G depletion without affecting its inhibitory activity.
Specifications
| APOBEC3G | |
| Polyclonal | |
| Unconjugated | |
| APOBEC3G | |
| A3G; APOBEC3G; APOBEC-related cytidine deaminase; APOBEC-related protein; APOBEC-related protein 9; apolipoprotein B editing enzyme catalytic polypeptide-like 3G; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3G; apolipoprotein B mRNA editing enzyme catalytic subunit 3G; apolipoprotein B mRNA editing enzyme cytidine deaminase; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3G; apolipoprotein B mRNA-editing enzyme catalytic polypeptide 3G; ARCD; ARP9; ARP-9; ASGPR2; ASGP-R2; bK150C2.7; CEM15; CEM-15; CLEC4H2; Deoxycytidine deaminase; dJ494G10.1; DNA dC->dU editing enzyme; DNA dC->dU-editing enzyme APOBEC-3G; FLJ12740; HBXBP; HL-2; MDS019; OTTHUMP00000028911; phorbolin-like protein MDS019 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 60489 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HC16 | |
| APOBEC3G | |
| Recombinant Protein corresonding to Human APOBEC3G. Recombinant protein control fragment (Product #RP-91476). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.