Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APOBEC3G Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS15407R

Description
The Apoliprotein B mRNA-editing, enzyme-catalytic, polypeptide-like (APOBEC) 3 is a multi-isoform member of the cytidine deaminase family of enzymes that act on monomeric nucleoside and nucleotide substrates. Similar to TRIM5-a which targets incoming retroviral capsids, APOBEC3 plays a major role in cellular defense against retroviral infection as at least two isoforms, APOBEC3G and to a lesser extent APOBEC3F, can be incorporated HIV-1 virions and induce hypermutation in the newly synthesized viral DNA and thus destabilize the viral genome. This innate mechanism of retroviral resistance is counteracted by the HIV-1 Vif protein by inducing the ubiquitization and degradation of APOBEC3G; a single amino acid substitution (D128K) blocks APOBEC3G depletion without affecting its inhibitory activity.Specifications
APOBEC3G | |
Polyclonal | |
Unconjugated | |
APOBEC3G | |
A3G; Apobec3; APOBEC3G; APOBEC-related cytidine deaminase; APOBEC-related protein; APOBEC-related protein 9; apolipoprotein B editing complex 3; apolipoprotein B editing enzyme catalytic polypeptide-like 3G; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3G; apolipoprotein B mRNA editing enzyme catalytic subunit 3G; apolipoprotein B mRNA editing enzyme cytidine deaminase; apolipoprotein B mRNA editing enzyme, catalytic polypeptide 3; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3G; Apolipoprotein B mRNA-editing complex 3; apolipoprotein B mRNA-editing enzyme catalytic polypeptide 3G; ARCD; Arp3; ARP9; ARP-9; ASGPR2; ASGP-R2; BC003314; bK150C2.7; CEM15; CEM-15; CLEC4H2; Deoxycytidine deaminase; dJ494G10.1; DNA dC->dU editing enzyme; DNA dC->dU-editing enzyme APOBEC-3; DNA dC->dU-editing enzyme APOBEC-3G; FLJ12740; HBXBP; HL-2; MDS019; OTTHUMP00000028911; phorbolin-like protein MDS019; recovery from Friend virus 3; Rfv3 | |
Rabbit | |
Protein A | |
RUO | |
60489 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9HC16 | |
APOBEC3G | |
KLH conjugated synthetic peptide derived from human APOBEC3G. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction