Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ APOBEC3H Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5139839
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: AETFRLQFNN KRRLRRPYYP RKALLCYQLT PQNGSTPTRG YFENKKKCHA EICFI.
DNA deaminase (cytidine deaminase) which acts as an inhibitor of retrovirus replication and retrotransposon mobility via deaminase-dependent and -independent mechanisms. The A3H-var/haplotype 2 exhibits antiviral activity against vif-deficient HIV-1. After the penetration of retroviral nucleocapsids into target cells of infection and the initiation of reverse transcription, it can induce the conversion of cytosine to uracil in the minus-sense single-strand viral DNA, leading to G-to-A hypermutations in the subsequent plus-strand viral DNA. The resultant detrimental levels of mutations in the proviral genome, along with a deamination-independent mechanism that works prior to the proviral integration, together exert efficient antiretroviral effects in infected target cells. Selectively targets single-stranded DNA and does not deaminate double-stranded DNA or single- or double-stranded RNA. Exhibits antiviral activity also against T-cell leukemia virus type 1 (HTLV-1) and may inhibit the mobility of LTR and non-LTR retrotransposons.
Specifications
| APOBEC3H | |
| Polyclonal | |
| Unconjugated | |
| APOBEC3H | |
| A3H; APOBEC3H; APOBEC-related protein 10; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3H; apolipoprotein B mRNA editing enzyme catalytic subunit 3H; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3H; Apolipoprotein B mRNA-editing enzyme catalytic polypeptide-like 3H; ARP10; ARP-10; DNA dC->dU-editing enzyme APOBEC-3H | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 164668 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6NTF7 | |
| APOBEC3H | |
| Recombinant protein corresponding to Human APOBEC3H. Recombinant protein control fragment (Product #RP-108962). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction