Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APOBEC3H Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554426
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554426 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554426 Supplier Invitrogen™ Supplier No. PA554426
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CSSCAWELVD FIKAHDHLNL GIFASRLYYH WCKPQQKGLR LLCGSQVPVE VMGFPEFADC WENFVDHEKP LSFNPYKMLE ELDKNSRAIK RRLERIKQS Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 51%.

DNA deaminase (cytidine deaminase) which acts as an inhibitor of retrovirus replication and retrotransposon mobility via deaminase-dependent and -independent mechanisms. The A3H-var/haplotype 2 exhibits antiviral activity against vif-deficient HIV-1. After the penetration of retroviral nucleocapsids into target cells of infection and the initiation of reverse transcription, it can induce the conversion of cytosine to uracil in the minus-sense single-strand viral DNA, leading to G-to-A hypermutations in the subsequent plus-strand viral DNA. The resultant detrimental levels of mutations in the proviral genome, along with a deamination-independent mechanism that works prior to the proviral integration, together exert efficient antiretroviral effects in infected target cells. Selectively targets single-stranded DNA and does not deaminate double-stranded DNA or single- or double-stranded RNA. Exhibits antiviral activity also against T-cell leukemia virus type 1 (HTLV-1) and may inhibit the mobility of LTR and non-LTR retrotransposons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APOBEC3H
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene APOBEC3H
Gene Accession No. Q6NTF7
Gene Alias A3H; APOBEC3H; APOBEC-related protein 10; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3H; apolipoprotein B mRNA editing enzyme catalytic subunit 3H; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3H; Apolipoprotein B mRNA-editing enzyme catalytic polypeptide-like 3H; ARP10; ARP-10; DNA dC->dU-editing enzyme APOBEC-3H
Gene Symbols APOBEC3H
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human APOBEC3H. Recombinant protein control fragment (Product #RP-92353).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 164668
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.