Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APOBR Monoclonal Antibody (OTI2E6), TrueMAB™, OriGene
SDP

Catalog No. 501681376
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-1376 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-1376 Supplier OriGene Supplier No. TA807047
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Macrophage receptor that binds to the apolipoprotein B48 (APOB) of dietary triglyceride (TG)-rich lipoproteins (TRL) or to a like domain of APOB in hypertriglyceridemic very low density lipoprotein (HTG-VLDL). Binds and internalizes TRL when out of the context of the macrophage. May provide essential lipids to reticuloendothelial cells. Could also be involved in foam cell formation with elevated TRL and remt lipoprotein (RLP). Mediates the rapid high-affinity uptake of chylomicrons (CM), HTG- VLDL, and trypsinized (tryp) VLDL devoid of APOE in vitro in macrophages.

Specifications

Antigen APOBR
Applications Western Blot
Classification Monoclonal
Clone OTI2E6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene APOBR
Gene Accession No. Q0VD83
Gene Alias APOB100R, APOB48R
Gene Symbols APOBR
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 780-1088 of human APOBR produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55911
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.