Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ApoC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577862
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577862 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577862 Supplier Invitrogen™ Supplier No. PA577862
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: human plasma Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Apolipoprotein C-III, also known as apo-CIII, is a protein that in humans is encoded by the APOC3 gene. Apo-CIII is a component of very low density lipoprotein (VLDL). APOC3 inhibits lipoprotein lipase and hepatic lipase; it is thought to delay catabolism of triglyceride-rich particles. The APOA1, APOC3 and APOA4 genes are closely linked in both rat and human genomes. The A-I and A-IV genes are transcribed from the same strand, while the A-1 and C-III genes are convergently transcribed. An increase in apoC-III levels induces the development of hypertriglyceridemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ApoC3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene APOC3
Gene Accession No. P02656
Gene Alias Apoc3; APOCIII; Apo-CIII; apoC-III; Apolipoprotein; Apolipoprotein C3; apolipoprotein C-3; apolipoprotein C-III; HALP2; MGC150353
Gene Symbols APOC3
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human Apolipoprotein CIII. The exact sequence is proprietary.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 345
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.