Learn More
APOE Mouse anti-Human, Clone: OTI6B9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF805276

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Apo E (Apolipoprotein E) plays an important role in the metabolism of lipids in the plasma, and is also is a constituent of various plasma lipoprotein-lipid particles. The protein is synthesized in liver, brain, spleen, and the kidney, and concentrations are high in interstitial fluids, where it plays a role in distributing cholesterol from cells where cholesterol is in excess to cells which require cholesterol. Apo E also plays an important role in the formation of very low density lipoprotein and chylomicrons. The Apo E gene is located on chromosome 19 at position 19q13.2.Specifications
APOE | |
Monoclonal | |
Unconjugated | |
APOE | |
AD2, APO-E, ApoE4, LDLCQ5, LPG | |
Mouse | |
Affinity Chromatography | |
RUO | |
348 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI6B9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P02649 | |
APOE | |
Full length human recombit protein of human APOE produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.