Learn More
APOE Polyclonal Antibody, Invitrogen™
Description
PA1-26902 detects Apolipoprotein E from human samples.
Specifications
Specifications
| Antigen | APOE |
| Applications | ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.125M sodium borate with 0.075M NaCl, 5mM EDTA and 0.01% sodium azide; pH 8 |
| Gene | APOE |
| Gene Accession No. | P02649, P02650 |
| Gene Alias | AD2; AI255918; APOE; apo-E; ApoE4; APOEA; Apolipoprotein; apolipoprotein E; apolipoprotein E3; apolipoprotein E4; Apoprotein; B2G1; BG; LDLCQ5; LPG; MGC1571 |
| Gene Symbols | APOE |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.