Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ApoE4 Antibody (4E4) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 3 publications
$206.50 - $501.50
Specifications
Antigen | ApoE4 |
---|---|
Clone | 40000 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:1000-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2-5 ug/ml, Immunoprecipitation 1:10-1:500 |
Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP149529SS
![]() |
Novus Biologicals
NBP149529SS |
0.025 mL |
Each for $206.50
|
|
|||||
NBP149529
![]() |
Novus Biologicals
NBP149529 |
0.1 mL |
Each for $501.50
|
|
|||||
Description
ApoE4 Monoclonal specifically detects ApoE4 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
ApoE4 | |
1 mg/mL | |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Alzheimers Research, Apoptosis, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
P02649 | |
348 | |
Synthetic peptide made to an internal region of human ApoE4 (within residues 100-150) [Uniprot: P02649] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
34 kDa |
40000 | |
Western Blot 1:1000-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2-5 ug/ml, Immunoprecipitation 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Apo-E, apolipoprotein E, apolipoprotein E3, late onset), MGC1571 | |
APOE | |
IgG1 κ | |
Protein G purified | |
This antibody is specific for ApoE4. It does not cross-react with the ApoE2 or ApoE3 isoforms. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title