Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ApoE4 Antibody (4E4), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP149529R
Description
ApoE4 Monoclonal specifically detects ApoE4 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
ApoE4 | |
Monoclonal | |
DyLight 550 | |
P02649 | |
APOE | |
Synthetic peptide made to an internal region of human ApoE4 (within residues 100-150) [Uniprot: P02649] | |
Protein G purified | |
RUO | |
Primary | |
This antibody is specific for ApoE4. It does not cross-react with the ApoE2 or ApoE3 isoforms. | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
40000 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
Apo-E, apolipoprotein E, apolipoprotein E3, late onset), MGC1571 | |
Mouse | |
34 kDa | |
0.1 mL | |
Alzheimers Research, Apoptosis, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
348 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction