Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ApoE4 Antibody (4E4), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP149529AF350
Description
ApoE4 Monoclonal specifically detects ApoE4 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
ApoE4 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
APOE | |
Synthetic peptide made to an internal region of human ApoE4 (within residues 100-150) [Uniprot: P02649] | |
0.1 mL | |
Primary | |
This antibody is specific for ApoE4. It does not cross-react with the ApoE2 or ApoE3 isoforms. | |
Store at 4°C in the dark. |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
40000 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
Apo-E, apolipoprotein E, apolipoprotein E3, late onset), MGC1571 | |
Mouse | |
Protein G purified | |
Alzheimers Research, Apoptosis, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
348.0 | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction