Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ ApoER2, Rat, Unlabeled, Clone: 4H3E6,

Catalog No. MABN1872 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN1872 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1872 Supplier MilliporeSigma™ Supplier No. MABN1872
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Low-density lipoprotein receptor-related protein 8 (UniProt Q924X6; also known as ApoER2, Apolipoprotein E receptor 2, LRP-8) is encoded by the Lrp8 (also known as Apoer2) gene (Gene ID 16975) in murine species. Murine ApoER2/LRP-8 is an 110kDa type I membrane receptor initially produced with a signal peptide sequence (a.a.1-28). The mature receptor has a large extracellular region (a.a. 29-858), a transmembrane domain (a.a. 859-881), and a cytoplasmic tail (a.a. 882-996). ApoER2 belongs to the LDLR family of membrane glycoproteins whose members also inculde the low-density lipoprotein receptor (LDLR), the very-low-density lipoprotein (VLDL) receptor, the LDLR-related protein (LRP), and Megalin (LRP-2). The overall extracellular structural features are similar among all LDLR family members. ApoER2 contains a cluster of eight cysteine-rich Complement-type/LDLR Class A repeats that mediate ligand-binding, followed by two EGF-like repeats (a.a. 364-408 and 409-448) and a cluster of five cysteine-poor LDLR Class B repeats (a.a. 495-714) that contains YWTD motifs involved in the pH-dependent ligands dissociation in endosomal compartments. ApoER2 and VLDLR both function as receptor for reelin, a glycoprotein that is mainly secreted by Cajal Retzius neurons in the marginal zone and plays an essential role in the formation of the layered neocortex. Immunohistochemical staining reveals differential localization of ApoER2 and VLDLR in the developing mouse cerebral cortex, with VLDLR being localized to the distal portion of leading processes in the marginal zone (MZ) and ApoER2 mainly localized to neuronal processes and the cell membranes of multipolar cells in the multipolar cell accumulation zone (MAZ). The differential expression pattern may therefore allow distinct reelin actions on migrating neurons during both the early and late migratory stages in the developing cerebral cortex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ApoER2
Applications Western Blot
Classification Monoclonal
Clone 4H3E6
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rat IgG2a in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene Lrp8, Apoer2
Gene Accession No. Q924X6
Host Species Rat
Immunogen GST-tagged recombinant mouse ApoER2 cytoplasmic tail lacking the 59-a.a. exon 19-coded sequence (Hirota, Y., et al. (2015). J. Comp. Neurol. 523(3):463-478).
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone 4H3E6 (3E) detected ApoER2 expression in embryonic and neonatal brain tissues from wild-type, but not Apoer2-knockout mice (Hirota, Y., et al. (2015). J. Comp. Neurol. 523(3):463-478).
Target Species Mouse
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.