Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ APOF Polyclonal Antibody
Rabbit Polyclonal Antibody
$178.40 - $511.50
Specifications
| Antigen | APOF |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
PIPA5118959
|
Invitrogen™
PA5118959 |
200 μL |
Each for $511.50
|
|
|||||
|
PIPA5122229
|
Invitrogen™
PA5122229 |
20 μL |
Each for $178.40
|
|
|||||
Description
APOF Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
The product of this gene is one of the minor apolipoproteins found in plasma. This protein forms complexes with lipoproteins and may be involved in transport and/or esterification of cholesterol.Specifications
| APOF | |
| -20°C | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| APOF | |
| AI255964; APOF; Apo-F; Apolipoprotein F; APON; Ba1-666; DKFZp781G18150; Leukemia virus-inactivating factor; lipid transfer inhibitor protein; Liver regeneration-related protein LRRG151; LTIP; LVIF; MGC22520; ovarian and testicular apolipoprotein N; Unknown (protein for MGC:137251) | |
| APOF | |
| Primary | |
| Antigen affinity chromatography |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300 | |
| Q13790 | |
| 319 | |
| Recombinant protein Apolipoprotein F. The antigen corresponds to amino acid range 122-307 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title