Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APOL1 Mouse anti-Human, Clone: 1G12D11, Proteintech
SDP

Catalog No. 50558984 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-558-984 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-558-984 Supplier Proteintech Group Inc Supplier No. 661241IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes the endogenous ApoL1 of 39-45 kDa in blood lysate. This antibody is specific to APOL1.

Apolipoproteins are protein components of plasma lipoproteins. The apolipoprotein L gene family encodes six highly homologous proteins designated apoL-I to -VI, which are associated with large high density type lipoproteins (HDL). The human apoL family maps to chromosome 22q12.1- 13.1 within a 127,000-bp region. apoL has been characterized as a pancreas specific, 383-amino acid, 42 kDa protein that contains a 12-amino acid secretory signal peptide. The apoL genes have TATA-less promoters and contain putative sterol regulatory elements, suggesting that transcription of these genes may be coordinated with that of the low density lipoprotein receptor and genes in pathways involving the synthesis of triglycerides and cholesterol. apoL homologs can undergo 10 fold changes in expression during atherosclerotic changes in vascular endothelial cells, which includes the inflammatory reaction of atherosclerotic lesions.

Specifications

Antigen APOL1
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone 1G12D11
Concentration 1.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene APOL1
Gene Accession No. O14791
Gene Alias APO L, APOL, APOL I, APOL1, Apolipoprotein L, Apolipoprotein L I, apolipoprotein L, 1, Apolipoprotein L1
Gene Symbols APOL1
Host Species Mouse
Immunogen APOL1-Specific Fusion Protein Ag2016
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8542
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.