Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APOL2 Monoclonal Antibody (C1)

Catalog No. PIMA543956
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543956 20 μL
PIMA541555 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543956 Supplier Invitrogen™ Supplier No. MA543956
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

APOL2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

This gene is a member of the apolipoprotein L gene family. The encoded protein is found in the cytoplasm, where it may affect the movement of lipids or allow the binding of lipids to organelles. Two transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APOL2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone C1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene APOL2
Gene Accession No. Q9BQE5
Gene Alias Apol2; ApoL-2; APOL3; Apol8; APOL-II; apolipoprotein L 8; apolipoprotein L, 2; Apolipoprotein L2; Apolipoprotein L-2; apolipoprotein L-II; OTTHUMP00000028936
Gene Symbols APOL2
Host Species Mouse
Immunogen Recombinant protein Apolipoprotein L2. The antigen corresponds to amino acid range 186-337 of the target protein.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23780, 315111
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.