Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APOL3 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP106979
Description
APOL3 Polyclonal specifically detects APOL3 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
APOL3 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
apoL-III, APOLIII, apolipoprotein L, 3, apolipoprotein L3, Apolipoprotein L-III, CG12_1, CG121, CG12-1, TNF-inducible protein CG12-1 | |
Goat | |
44 kDa | |
0.1 mg | |
Signal Transduction | |
80833 | |
Human | |
IgG |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:1000 | |
NP_663615.1 | |
APOL3 | |
Peptide with sequence ELTQIYQRLNPCHTH corresponding to C-Terminus according to NP_663615.1, NP_055164.1, NP_663616.1. | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is expected to recognize isoform 1 (NP_663615.1), isoform 2 (NP_055164.1, NP_085147.1, NP_663614.1) and isoform 3 (NP_663616.1, NP_663617.1). | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction