Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APOLD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP179330
Description
APOLD1 Polyclonal specifically detects APOLD1 in Human samples. It is validated for Western Blot.Specifications
APOLD1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
apolipoprotein L domain containing 1, apolipoprotein L domain-containing protein 1, DKFZP434F0318, FLJ25138, FLJ95166, Vascular early response gene protein, VERGEDKFZp434F0318 | |
Rabbit | |
30 kDa | |
100 μL | |
Primary | |
Centrifuge vial prior to reconstitution. Add 50μL distilled water to a final antibody concentration of 1mg/mL. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q96LR9 | |
APOLD1 | |
Synthetic peptide directed towards the middle region of human APOLD1The immunogen for this antibody is APOLD1. Peptide sequence LIPRRAEGDTKVSQAVLKAKIQKLAESLESCTGALDELSEQLESRVQLCT. | |
Affinity purified | |
RUO | |
81575 | |
Human, Mouse, Rat, Bovine, Canine, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction