Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APOLD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562616
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562616 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562616 Supplier Invitrogen™ Supplier No. PA562616
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLIFCNSREL RRVQEIAATC QDQMREILSC LEFFCRWQGC GDRQLLQCGR NASI Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 87%.

May be involved in angiogenesis. May play a role in activity-dependent changes of brain vasculature. May affect blood-brain permeability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APOLD1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene APOLD1
Gene Accession No. Q96LR9
Gene Alias APOLD1; apolipoprotein L domain containing 1; apolipoprotein L domain-containing protein 1; Vascular early response gene protein; VERGE
Gene Symbols APOLD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human APOLD1. Recombinant protein control fragment (Product #RP-97864).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81575
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.