Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein A-I/ApoA1 Antibody (003), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289598APC
Description
Apolipoprotein A-I/ApoA1 Monoclonal antibody specifically detects Apolipoprotein A-I/ApoA1 in Human samples. It is validated for ELISASpecifications
Apolipoprotein A-I/ApoA1 | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
003 | |
ELISA | |
APOA1, Apo-AI, ApoA-I, Apolipoprotein A1, apolipoprotein A-I, MGC117399 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Apolipoprotein A-I/ApoA1 (Uniprot#: P02647; Asp25-Gln267). | |
0.1 mL | |
Biologically Active Proteins, Breast Cancer, Cancer, Diabetes Research, Lipid and Metabolism | |
335 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction