Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein A-I/ApoA1 Antibody (1E12) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197943100UL
Description
Apolipoprotein A-I/ApoA1 Monoclonal specifically detects Apolipoprotein A-I/ApoA1 in Human samples. It is validated for Western Blot, ELISA.Specifications
Apolipoprotein A-I/ApoA1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:5000, Flow Cytometry, ELISA 1:100-1:2000 | |
NP_000030.1 | |
APOA1 | |
Recombinant human Apolipoprotein A1 (25-267aa) purified from E. coli | |
100 μL | |
Breast Cancer, Cancer, Diabetes Research, Lipid and Metabolism | |
335 | |
Human | |
Purified |
Western Blot, Flow Cytometry, ELISA | |
1000000000000 | |
Unconjugated | |
PBS (pH 7.4) and 10% Glycerol with 0.02% Sodium Azide | |
APOA1, Apo-AI, ApoA-I, Apolipoprotein A1, apolipoprotein A-I, MGC117399 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human Apolipoprotein A1 mAb, clone 1E12, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human APOA1 protein. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction