Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein A-I/ApoA1 Antibody (1E12), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197943F
Description
Apolipoprotein A-I/ApoA1 Monoclonal specifically detects Apolipoprotein A-I/ApoA1 in Human samples. It is validated for ELISA.Specifications
Apolipoprotein A-I/ApoA1 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
APOA1 | |
Recombinant human Apolipoprotein A1 (25-267aa) purified from E. coli | |
100 μL | |
Biologically Active Proteins, Breast Cancer, Cancer, Diabetes Research, Lipid and Metabolism | |
335 | |
Human | |
Purified |
ELISA | |
1000000000000 | |
ELISA | |
APOA1, apoA-I, Apo-AI, Apolipoprotein A1, apolipoprotein A-I, MGC117399 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human Apolipoprotein A1 mAb, clone 1E12, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human APOA1 protein. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction