Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein A-I/ApoA1 Antibody (2083A), DyLight 680, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB36641Y
Description
Apolipoprotein A-I/ApoA1 Monoclonal antibody specifically detects Apolipoprotein A-I/ApoA1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry (Intracellular Staining), ImmunocytochemistrySpecifications
Apolipoprotein A-I/ApoA1 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry | |
2083A | |
Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry | |
APOA1, Apo-AI, ApoA-I, Apolipoprotein A1, apolipoprotein A-I, MGC117399 | |
E. coli-derived recombinant human ApoA1, Asp25-Gln267, Accession # P02647 | |
0.1 mL | |
Biologically Active Proteins, Breast Cancer, Cancer, Diabetes Research, Lipid and Metabolism | |
335 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction