Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein A-I/ApoA1 Antibody (2083D), Alexa Fluor 532, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals MAB3664AF532
Description
Apolipoprotein A-I/ApoA1 Monoclonal specifically detects Apolipoprotein A-I/ApoA1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry.Specifications
Apolipoprotein A-I/ApoA1 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
APOA1 | |
rhApoA1 aa25-267 | |
0.1 mL | |
Primary | |
Human | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry | |
2083D | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry | |
APOA1, apoA-I, Apo-AI, Apolipoprotein A1, apolipoprotein A-I, MGC117399 | |
Rabbit | |
Protein A or G purified | |
Biologically Active Proteins, Breast Cancer, Cancer, Diabetes Research, Lipid and Metabolism | |
335.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction