Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein A-I/ApoA1 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP252979C
Description
Apolipoprotein A-I/ApoA1 Polyclonal specifically detects Apolipoprotein A-I/ApoA1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Apolipoprotein A-I/ApoA1 | |
Polyclonal | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
APOA1, apoA-I, Apo-AI, Apolipoprotein A1, apolipoprotein A-I, MGC117399 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
APOA1 | |
Partial recombinant human ApoA1 protein (amino acids 25-267) [UniProt P02647] | |
0.1 mL | |
Biologically Active Proteins, Breast Cancer, Cancer, Diabetes Research, Lipid and Metabolism | |
335 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction