Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein A-I Mouse anti-Human, Unconjugated, Clone: Wt20-7, MilliporeSigma™

Catalog No. MABS1310100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS1310-100 100 μL
MABS1310-25 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABS1310-100 Supplier MilliporeSigma Supplier No. MABS1310100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Apolipoprotein A-I, clone Wt20-7, Cat. No. MABS1310, is a mouse monoclonal antibody that detects human Apolipoprotein A-I and has been tested for use in Dot Blot, ELISA, Immunocytochemistry, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Apolipoprotein A-I
Applications Dot Blot, ELISA, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone Wt20-7
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Apo-AI;ApoA-I;Apolipoprotein A1
Gene Symbols APOA1
Host Species Mouse
Immunogen GST-tagged full-length human recombinant Apolipoprotein A-I
Molecular Weight of Antigen ∽28 kDa observed; 30.78 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_000030
Test Specificity Clone Wt20-7 specifically detects human Apolipoprotein A-I.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.