Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Apolipoprotein A1 ELISA Kit

Catalog No. p-7230927
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHAPOA1 96 Tests
EHAPOA1X10 10 x 96 Tests
EHAPOA1X5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EHAPOA1 Supplier Invitrogen™ Supplier No. EHAPOA1
Only null left
Add to Cart
Add to Cart

ELISA

The Human Apolipoprotein A1 (Hu ApoA1) ELISA quantitates Hu ApoA1 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu ApoA1. Principle of the method The Human ApoA1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Apolipoprotein A-1 (apo A-1) in human plasma is a major protein component of high density lipoprotein (HDL). The molecular mass is 24 kDa. Apo A-1 is a cofactor for enzymes involved in the normal turnover of cholesterol and is produced in the liver. Plasma concentration is 270-380 mg/100 mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P02647
Assay Range 0.082 to 20 ng/mL
Assay Sensitivity 0.08 ng/mL
Calibration Range 0.08 ng/mL to 20 ng/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Product Type ELISA
Protein Family GCPR
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA pair antibody detects human ApoA1. Cross-reacts with ApoA1 from rhesus and cynomolgus macaques. Other species not determined.
Sufficient For 96 assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 335
Gene Symbol apo(a)
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Cardiovascular Research
Sample Volume Plasma 1 μL, Serum 1 μL, Supernatant 100 μL
Storage Requirements 2°C to 8°C
Synonym N/A
Target ApoA1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.