Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein A5 Antibody (4H8H8E2), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11057307AF350
Description
Apolipoprotein A5 Monoclonal specifically detects Apolipoprotein A5 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Apolipoprotein A5 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
APOA5 | |
Ni-NTA purified full-length recombinant human Apolipoprotein A5 expressed in E. Coli strain BL21 (DE3). [UniProtNo. Q6Q788] | |
0.1 mL | |
Primary | |
Human | |
IgG1 κ |
Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
4H8H8E2 | |
Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
Apo-AV, ApoA-V, APOAV, Apolipoprotein A5, apolipoprotein A-V, FLJ97995, MGC126836, RAP3MGC126838, Regeneration-associated protein 3 | |
Mouse | |
Protein G purified | |
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
116519.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction