Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Apolipoprotein A5 Monoclonal Antibody (1G5G9)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA116809
Description
The target sequence has 83% sequence homology with bovine, 79% sequence homology with rat and 78% sequence homology with mouse.
Apolipoprotein A4 (also known as ApoA-IV) is a plasma protein that is O-linked glycoprotein after proteolytic processing. ApoA4 is a major constituent of HDL and it also can be found on the surface of newly synthesized chylomicrons. The physiological function of ApoA4 is not completely identified so far, but several evidences from vitro studies indicate that this apolipoprotein involves in several steps of the reverse cholesterol transport pathway, which transports cholesterol from peripheral cells back to the liver.
Specifications
Apolipoprotein A5 | |
Monoclonal | |
Conc. Not Determined | |
Ascites with 0.1% sodium azide | |
Q6Q788 | |
APOA5 | |
Ni-NTA purified truncated recombinant human APOA5 expressed in E. Coli strain BL21 (DE3). | |
RUO | |
100772884, 116519 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1G5G9 | |
Unconjugated | |
APOA5 | |
1300007O05Rik; APOA5; Apoav; apo-AV; apoA-V; apolipoprotein A5; apolipoprotein AV; Apolipoprotein A-V; apolipoprotein A-V precursor variant 3; apolipoprotein A-V; apolipoprotein A5; FLJ97995; MGC126836; MGC126838; Rap3; Regeneration-associated protein 3; UNQ411/PRO773 | |
Mouse | |
100 μL | |
Primary | |
Human, Hamster | |
Antibody | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction