Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Apolipoprotein A5 Monoclonal Antibody (2G1H11, 1F1E8)
GREENER_CHOICE

Catalog No. PIMA515265
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515265 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515265 Supplier Invitrogen™ Supplier No. MA515265
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15265 targets APOA5 in indirect ELISA, IF and WB applications and shows reactivity with Human samples. The MA5-15265 immunogen is purified recombinant fragment of human Apoa5 expressed in E. Coli..

Apolipoprotein A4 (also known as ApoA-IV) is a plasma protein that is O-linked glycoprotein after proteolytic processing. ApoA4 is a major constituent of HDL and it also can be found on the surface of newly synthesized chylomicrons. The physiological function of ApoA4 is not completely identified so far, but several evidences from vitro studies indicate that this apolipoprotein involves in several steps of the reverse cholesterol transport pathway, which transports cholesterol from peripheral cells back to the liver.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Apolipoprotein A5
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2G1H11, 1F1E8, 1F1E8
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene APOA5
Gene Accession No. Q6Q788
Gene Alias 1300007O05Rik; APOA5; Apoav; apo-AV; apoA-V; apolipoprotein A5; apolipoprotein AV; Apolipoprotein A-V; apolipoprotein A-V precursor variant 3; apolipoprotein A-V; apolipoprotein A5; FLJ97995; MGC126836; MGC126838; Rap3; Regeneration-associated protein 3; UNQ411/PRO773
Gene Symbols APOA5
Host Species Mouse
Immunogen Purified recombinant fragment of human Apoa5 expressed in E. Coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116519
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.