Learn More
Invitrogen™ Apolipoprotein A5 Monoclonal Antibody (4H8H8E2)

Description
Suggested positive control: antigen standard for APOA5 (transient overexpression lysate).
Specifications
Specifications
| Antigen | Apolipoprotein A5 |
| Applications | ELISA, Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 4H8H8E2 |
| Concentration | 1.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | APOA5 |
| Gene Accession No. | Q6Q788 |
| Gene Alias | 1300007O05Rik; APOA5; Apoav; apo-AV; apoA-V; apolipoprotein A5; apolipoprotein AV; Apolipoprotein A-V; apolipoprotein A-V precursor variant 3; apolipoprotein A-V; apolipoprotein A5; FLJ97995; MGC126836; MGC126838; Rap3; Regeneration-associated protein 3; UNQ411/PRO773 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.