Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein A5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101429
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101429 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101429 Supplier Thermo Scientific Supplier No. PA5101429
Add to Cart
Add to Cart

Apolipoprotein A5 Polyclonal Antibody

Apolipoprotein A5 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen Apolipoprotein A5
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene APOA5
Gene Accession No. Q6Q788
Gene Alias 1300007O05Rik; APOA5; Apoav; apo-AV; apoA-V; apolipoprotein A5; apolipoprotein AV; Apolipoprotein A-V; apolipoprotein A-V precursor variant 3; apolipoprotein A-V; apolipoprotein A5; FLJ97995; MGC126836; MGC126838; Rap3; Regeneration-associated protein 3; UNQ411/PRO773
Gene Symbols APOA5
Host Species Rabbit
Immunogen A synthesized peptide derived from human APOA5(Accession Q6Q788), corresponding to amino acid residues A13-D63.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116519
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.