Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein B/ApoB Antibody (APOB/3300), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308226G
Description
Apolipoprotein B/ApoB Monoclonal antibody specifically detects Apolipoprotein B/ApoB in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein ArraySpecifications
Apolipoprotein B/ApoB | |
Monoclonal | |
DyLight 488 | |
Apo B-100, apoB-100, apoB-48, apolipoprotein B (including Ag(x) antigen), apolipoprotein B-100, apolipoprotein B48, FLDB, LDLCQ4, mutant Apo B 100 | |
Human recombinant Apolipoprotein B/ApoB protein fragment (around aa592-689) (exact sequence is proprietary) (Uniprot: P04114) | |
100 μg | |
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
338 | |
Store at 4°C in the dark. | |
IgG2b κ |
Immunohistochemistry (Paraffin), Protein Array | |
APOB/3300 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction