Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein B/ApoB Antibody (SR1898), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32171450UL
Description
Apolipoprotein B/ApoB Monoclonal antibody specifically detects Apolipoprotein B/ApoB in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunoprecipitationSpecifications
Apolipoprotein B/ApoB | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunoprecipitation | |
SR1898 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunoprecipitation 1:50 | |
Apo B-100, apoB-100, apoB-48, apolipoprotein B (including Ag(x) antigen), apolipoprotein B-100, apolipoprotein B48, FLDB, LDLCQ4, mutant Apo B 100 | |
A synthesized peptide derived from human Apolipoprotein B/ApoB (Uniprot #: P04114) | |
50 μg | |
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
338 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction