Learn More
Invitrogen™ Apolipoprotein B Polyclonal Antibody

Description
Immunogen sequence: NFVASHIANI LNSEELDIQD LKKLVKEALK ESQLPTVMDF RKFSRNYQLY KSVSLPSLDP ASAKIEGNLI FDPNNYLPKE SMLKTTLTAF GFASADLIE Highest antigen sequence indentity to the following orthologs: Rat - 78%, Mouse - 76%.
Specifications
Specifications
| Antigen | Apolipoprotein B |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | APOB |
| Gene Accession No. | P04114 |
| Gene Alias | Aa1064; Ac1-060; AI315052; Apo B100; Apo B-100; Apo B-48; APOB; apo-B; ApoB 100; ApoB 48; ApoB-100; apob-48; apolipo b; apolipoprotein B; apolipoprotein B (including Ag(x) antigen); Apolipoprotein B 100; Apolipoprotein B 48; apolipoprotein B PI; Apolipoprotein B100; apolipoprotein B-100; apolipoprotein B46; apolipoprotein B47; apolipoprotein B48; Apolipoprotein B-48; apolipoprotein B49; FLDB; LDLCQ4; LOX-1; mCG_129875; MGC176318; Ox-LDL receptor 1 |
| Gene Symbols | APOB |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.