Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Apolipoprotein C1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111339
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111339 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111339 Supplier Invitrogen™ Supplier No. PA5111339
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PDVSSALDKL KEFGNTLEDK ARELISRIKQ SELSAKMREW FSETFQKVKE Highest antigen sequence indentity to the following orthologs: Mouse - 65%, Rat - 65%.

Apolipoproteins are protein components of plasma lipoproteins. The apolipoprotein C gene family encodes four homologous proteins designated apoC-I to -IV, which specifically modulate the metabolism of triglyceride-rich lipoproteins. The human apoC-I gene maps to chromosome 19q13.2 and is expressed primarily in the liver where it is activated when monocytes differentiate into macrophages. The human apoC-II gene maps to chromosome 19q13.2 and encodes a 79 amino acid single chain protein that is a necessary cofactor for the activation of lipoprotein lipase, the enzyme that hydrolyzes triglycerides in plasma and transfers the fatty acids to tissues. The human apoC-III gene maps to chromosome 11q23 and encodes a protein that may delay catabolism of triglyceride-rich particles by inhibiting lipoprotein lipase and hepatic lipase. The human apoC-IV gene maps to chromosome 19q13.2 and encodes a 127 amino acid protein that is primarily expressed in the liver.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Apolipoprotein C1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene APOC1
Gene Accession No. P02654
Gene Alias ALPCI; Apoc1; Apo-CI; ApoC-I; Apo-CIB; ApoC-IB; apolipoprotein C1; Apolipoprotein C-I; apolipoprotein C-I variant I; apolipoprotein C-I variant II; Liver regeneration-related protein LRRG04; LRRG04; Truncated apolipoprotein C-I
Gene Symbols APOC1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human Apolipoprotein C1. Recombinant protein control fragment (Product #RP-101269).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 341
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.