Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein CIII Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB600610
Description
Apolipoprotein CIII Polyclonal specifically detects Apolipoprotein CIII in Human, Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
Apolipoprotein CIII | |
Polyclonal | |
Unconjugated | |
0.125 M Sodium Borate, 0.075 M Sodium Chloride, 0.005 M EDTA, pH 8.0 with 0.01% Sodium Azide | |
APOC3, apoC-III, Apo-CIII, APOCIII, Apolipoprotein C3, apolipoprotein C-III, MGC150353 | |
Goat | |
10.85 kDa | |
0.1 mg | |
Primary | |
Typically less than 1% cross reactivity against other types of apoLipoprotein was detected by ELISA against purified standards. This antibody reacts with human apoLipoprotein C-III and has negligible cross-reactivity with Type A-I, A-II, B, C-I, C-II, E and J apoLipoproteins. Specific cross reaction of anti-apoLipoprotein antibodies with antigens from other species has not been determined. Non-specific cross reaction of anti-apoLipoprotein antibodies with other human serum proteins is negligible. | |
Store at 4°C short term. For extended storage, add an equal volume of glycerol, aliquot and store at -20°C or below. Avoid repeated freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
1 mg/ml | |
Western Blot 1:500-1:5000, ELISA 1:5000-1:10000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:10-1:500 | |
P02656 | |
APOC3 | |
produced from Apolipoprotein CIIII derived from human plasma (Uniprot: P02656) | |
Affinity Purified | |
RUO | |
345 | |
Human, Bovine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction