Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein D Antibody (APOD/3415), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314090AF647
This item is not returnable.
View return policy
Description
Apolipoprotein D Monoclonal antibody specifically detects Apolipoprotein D in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Apolipoprotein D | |
| Monoclonal | |
| Alexa Fluor 647 | |
| apo-D, ApoD, apolipoprotein D | |
| Human recombinant Apolipoprotein D protein fragment (around aa21-119) (exact sequence is proprietary) (Uniprot: P05090) | |
| 0.1 mL | |
| Alzheimers Research, Cancer, Cardiovascular Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 347 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| APOD/3415 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction