Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein D Antibody (SR1458), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322198100UL
Description
Apolipoprotein D Monoclonal antibody specifically detects Apolipoprotein D in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
Apolipoprotein D | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
SR1458 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
apo-D, ApoD, apolipoprotein D | |
A synthesized peptide derived from human Apolipoprotein D (Uniprot #: P05090) | |
100 μg | |
Alzheimers Research, Cancer, Cardiovascular Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
347 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction