Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein D Mouse anti-Human, Clone: D544, Novus Biologicals™

Mouse Monoclonal Antibody
$286.00 - $1033.00
Specifications
Antigen | Apolipoprotein D |
---|---|
Clone | D544 |
Concentration | 0.5 mg/ml |
Applications | ELISA |
Classification | Monoclonal |
Description
Apolipoprotein D Monoclonal antibody specifically detects Apolipoprotein D in Human samples. It is validated for ELISASpecifications
Apolipoprotein D | |
0.5 mg/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
apo-D, ApoD, apolipoprotein D | |
Recombinant human Apolipoprotein D | |
Primary | |
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
D544 | |
ELISA | |
Unconjugated | |
Mouse | |
Alzheimers Research, Cancer, Cardiovascular Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
0.2 um filtered PBS | |
347 | |
IgG1 | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title