Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein E/ApoE Antibody (WUE-4), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11060531AF647
Description
Apolipoprotein E/ApoE Monoclonal specifically detects Apolipoprotein E/ApoE in Human, Mouse, Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).Specifications
Apolipoprotein E/ApoE | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Apo-E, apolipoprotein E, apolipoprotein E3, late onset), MGC1571 | |
Mouse | |
36 kDa | |
0.1 mL | |
Alzheimers Research, Apoptosis, Cancer, Cholesterol Metabolism, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
348 | |
Human, Mouse, Rat (Negative) | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
WUE-4 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular) | |
P02649 | |
APOE | |
Purified human ApoE [UniProt# P02649] | |
Protein G purified | |
RUO | |
Primary | |
This antibody detects ApoE2, ApoE3 and ApoE4 (PMID 2466929). | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction