Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein E/ApoE Antibody (WUE-4), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11060531APC
Description
Apolipoprotein E/ApoE Monoclonal specifically detects Apolipoprotein E/ApoE in Human, Mouse, Rat (Negative) samples. It is validated for Flow Cytometry, Flow (Intracellular).Specifications
Apolipoprotein E/ApoE | |
Monoclonal | |
APC | |
PBS with 0.05% Sodium Azide | |
APOE | |
Purified human ApoE [UniProt# P02649] | |
0.1 mL | |
Primary | |
This antibody detects ApoE2, ApoE3 and ApoE4 (PMID 2466929). | |
Store at 4C in the dark. | |
IgG1 κ |
Flow Cytometry | |
WUE-4 | |
Flow Cytometry, Flow (Intracellular) | |
Apo-E, apolipoprotein E, apolipoprotein E3, late onset), MGC1571 | |
Mouse | |
Protein G purified | |
Alzheimers Research, Apoptosis, Cancer, Cholesterol Metabolism, Cognition and Behavior, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
348 | |
Human, Mouse, Rat (Negative) | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction