Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein E R2/ApoE R2 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002217MFV610
Description
Apolipoprotein E R2/ApoE R2 Polyclonal antibody specifically detects Apolipoprotein E R2/ApoE R2 in Human, Mouse, Bovine, Chicken samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
Apolipoprotein E R2/ApoE R2 | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence | |
APOER2HSZ75190, Apolipoprotein E receptor 2, low density lipoprotein receptor-related protein 8, apolipoprotein e receptor, low-density lipoprotein receptor-related protein 8, LRP-8, MCI1 | |
A synthetic peptide made to a C-terminal portion of the human ApoER2 protein sequence (between residues 800-900). [UniProt# Q14114] | |
0.1 mL | |
Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
7804 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Bovine, Chicken | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction