Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein E R2/ApoE R2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $499.50

Specifications

Antigen Apolipoprotein E R2/ApoE R2
Concentration 0.96 mg/mL
Dilution Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 5 ug/ml
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB1002217SS
SDP
View Documents
Novus Biologicals
NB1002217SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NB1002217
SDP
View Documents
Novus Biologicals
NB1002217
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

Apolipoprotein E R2/ApoE R2 Polyclonal specifically detects Apolipoprotein E R2/ApoE R2 in Human, Mouse, Bovine, Chicken samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Apolipoprotein E R2/ApoE R2
Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 5 ug/ml
Polyclonal
Rabbit
Alzheimers Research, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction
Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide
APOER2HSZ75190, Apolipoprotein E receptor 2, low density lipoprotein receptor-related protein 8, apolipoprotein e receptor, low-density lipoprotein receptor-related protein 8, LRP-8, MCI1
LRP8
IgG
Affinity Purified
0.96 mg/mL
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
Human, Mouse, Bovine, Chicken
Q14114
7804
A synthetic peptide made to a C-terminal portion of the human ApoER2 protein sequence (between residues 800-900). [UniProt# Q14114]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.