Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein E3/ApoE3 Antibody (960319), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals MAB41442F
Description
Apolipoprotein E3/ApoE3 Monoclonal specifically detects Apolipoprotein E3/ApoE3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Apolipoprotein E3/ApoE3 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
APOE | |
ApoE3 variant peptide found within amino acids 100-200. | |
100 μL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
960319 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
AD2, Apo-E, ApoE4, apolipoprotein E, LDLCQ5, LPG | |
Mouse | |
Protein A or G purified | |
RUO | |
348 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction