Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Apolipoprotein E4 Human ELISA Kit

Catalog No. EHAPOE4X5
Encompass_Preferred
Click to view available options
Quantity:
5 x 96 Tests
96 Tests

Sandwich ELISA Kit

The Human Apolipoprotein E4 (Hu ApoE4) ELISA quantitates Hu ApoE4 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu ApoE4. Principle of the method The Human ApoE4 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Apo E (Apolipoprotein E) plays an important role in the metabolism of lipids in the plasma, and is also is a constituent of various plasma lipoprotein-lipid particles. The protein is synthesized in liver, brain, spleen, and the kidney, and concentrations are high in interstitial fluids, where it plays a role in distributing cholesterol from cells where cholesterol is in excess to cells which require cholesterol. Apo E also plays an important role in the formation of very low density lipoprotein and chylomicrons. The Apo E gene is located on chromosome 19 at position 19q13.2.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P02649
Assay Range 0.41 ng/mL to 100 ng/mL
Assay Sensitivity 0.4 ng/mL
Calibration Range 0.4 ng/mL to 100 ng/mL
Assay Solid-phase sandwich ELISA
Conjugate Biotin
Product Type ELISA Kit
Protein Family GCPR
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA pair antibody detects human ApoE4. Other species not determined.
Sufficient For 5 x 96 assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 348
Gene Symbol AD2,APO-E,ApoE4,LDLCQ5,LPG
Interassay CV <12%
Intraassay CV <10%
Kit Contents Stop Solution, SAV-HRP, Assay Diluent Concentrate, Biotinylated Detection Antibody, Wash Buffer, Pre-coated 96 well plates, Adhesive Plate Covers, Chromogen, Standard
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Research Discipline Cardiovascular Research
Sample Volume Plasma, 1 μ;L; Serum, 1 μ;L; Supernatant, 100 μ;L
Storage Requirements 2°C to 8°C
Synonym AD2, APO-E, LDLCQ5, LPG
Target Human
Test Time 1 hr. 20 min.
Total Assay Time 4 hr. 45 min.
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.