Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apolipoprotein F Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299242APC
Description
Apolipoprotein F Polyclonal antibody specifically detects Apolipoprotein F in Mouse samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Apolipoprotein F | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
Apo-F, apolipoprotein F, DKFZp781G18150, Lipid transfer inhibitor protein, LTIP, MGC22520 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Mouse Apolipoprotein F. | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
319 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction